Learn More
Apexbio Technology LLC Wnt-C59, 1243243-89-1, MFCD22201167, 5 mg
SDP
Supplier:  Apexbio Technology LLC A86855
Encompass_Preferred
MF: C25H21N3O, MW: 379.45, Solubility: 18.95mg/mL in DMSO. Wnt-C59 is a selective inhibitor of Wnt signaling with IC50 value of 74 pM.
Catalog No. 50-101-4716