Learn More
Selleck Chemical LLC Wnt-C59 (C59) 5mg 1243243-89-1
SDP
Supplier:  Selleck Chemical LLC S70375MG
Encompass_Preferred
Wnt-C59 (C59) is a PORCN inhibitor for Wnt3A-mediated activation of a multimerized TCF-binding site driving luciferase with IC50 of 74 pM in HEK293 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0710557