Learn More
Medchemexpress LLC Wnt/β-catenin agonist 2 | 943820-93-7 | 99.7% | C13H12N4O3 | 100 MG
SDP
Catalog No. 5000354676
Encompass_Preferred
Wnt/β-catenin agonist 2 is a potent Wnt agonist that activates Wnt/β-catenin signaling. It is used in research related to signal transduction diseases. This compound is an activator for β-Catenin in the SW480 cell line and affects the differentiation of ST2 cells into osteoblasts.
  • Potent Wnt agonist
  • Activates Wnt/β-catenin signaling
  • Used in research for signal transduction diseases
  • Activator for β-Catenin in SW480 cell line
  • Affects differentiation of ST2 cells into osteoblasts
Catalog No. 50-003-54676 Supplier Medchemexpress LLC Supplier No. HY141873100MG
Wnt/β-catenin agonist 2 is a potent Wnt agonist that activates Wnt/β-catenin signaling. It is used in research related to signal transduction diseases. This compound is an activator for β-Catenin in the SW480 cell line and affects the differentiation of ST2 cells into osteoblasts.
  • Potent Wnt agonist
  • Activates Wnt/β-catenin signaling
  • Used in research for signal transduction diseases
  • Activator for β-Catenin in SW480 cell line
  • Affects differentiation of ST2 cells into osteoblasts