Learn More
Medchemexpress LLC Wnt pathway inhibitor 2 | 1144043-83-3 | 99.8% | 703.59 | 100 MG
SDP
Catalog No. 5000348711
Encompass_Preferred
YB-0158 (Wnt pathway inhibitor 2) is a reverse-turn peptidomimetic and a potent colorectal cancer stem cell (CSC) targeting agent. It disrupts Sam68-Src interactions and induces apoptosis in CRC cells, demonstrating anti-cancer activities. In vitro studies have shown it significantly increases apoptosis in CRC cells and decreases CBP recruitment at the promoter of Wnt/beta-Catenin target genes.
  • Reverse-turn peptidomimetic
  • Potent colorectal cancer stem cell (CSC) targeting agent
  • Disrupts Sam68-Src interactions
  • Induces apoptosis in CRC cells
  • Demonstrates anti-cancer activities
  • Increases apoptosis in CRC cells (in vitro)
  • Decreases CBP recruitment at the promoter of Wnt/beta-Catenin target genes (in vitro)
  • For research use only
Catalog No. 50-003-48711 Supplier Medchemexpress LLC Supplier No. HY136541100MG
YB-0158 (Wnt pathway inhibitor 2) is a reverse-turn peptidomimetic and a potent colorectal cancer stem cell (CSC) targeting agent. It disrupts Sam68-Src interactions and induces apoptosis in CRC cells, demonstrating anti-cancer activities. In vitro studies have shown it significantly increases apoptosis in CRC cells and decreases CBP recruitment at the promoter of Wnt/beta-Catenin target genes.
  • Reverse-turn peptidomimetic
  • Potent colorectal cancer stem cell (CSC) targeting agent
  • Disrupts Sam68-Src interactions
  • Induces apoptosis in CRC cells
  • Demonstrates anti-cancer activities
  • Increases apoptosis in CRC cells (in vitro)
  • Decreases CBP recruitment at the promoter of Wnt/beta-Catenin target genes (in vitro)
  • For research use only