Learn More
TARGETMOL CHEMICALS INC WNT PATHWAY INHIBITOR 3 5MG
SDP
Catalog No. 502579142
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Wnt pathway inhibitor 3 is a potent AC1 inhibitor with an IC50 value of 45 nM.Wnt pathway inhibitor 3 has antiproliferative activity and can be used in studies to ameliorate osteoarthritis in a mouse model of experimental osteoarthritis. purity: 99%
Catalog No. 50-257-9142 Supplier TARGETMOL CHEMICALS INC Supplier No. T775025MG
Also available in 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Wnt pathway inhibitor 3 is a potent AC1 inhibitor with an IC50 value of 45 nM.Wnt pathway inhibitor 3 has antiproliferative activity and can be used in studies to ameliorate osteoarthritis in a mouse model of experimental osteoarthritis. purity: 99%