Learn More
Medchemexpress LLC Wnt pathway inhibitor 3 | 663213-98-7 | C21H17BrN2O5 | 5 MG
SDP
Catalog No. 5000376026
Encompass_Preferred
Wnt pathway inhibitor 3 is a potent Wnt inhibitor with an IC50 value of 45 nM. It exhibits antiproliferative activity. This product is for research use only.
  • Potent Wnt inhibitor with an IC50 value of 45 nM
  • Exhibits antiproliferative activity
  • IC50 values for HS68, Dld1, HCT116, and SW480 cells are 641, 470, 551, and 618 nM, respectively
  • For research use only
Catalog No. 50-003-76026 Supplier Medchemexpress LLC Supplier No. HY1537505MG
Add to Cart
Add to Cart
Wnt pathway inhibitor 3 is a potent Wnt inhibitor with an IC50 value of 45 nM. It exhibits antiproliferative activity. This product is for research use only.
  • Potent Wnt inhibitor with an IC50 value of 45 nM
  • Exhibits antiproliferative activity
  • IC50 values for HS68, Dld1, HCT116, and SW480 cells are 641, 470, 551, and 618 nM, respectively
  • For research use only