Learn More
Enzo Life Sciences SCREEN-WELL Wnt Pathway library (500 µl/well)
SDP
Catalog No. 502010216
Encompass_Preferred
The SCREEN-WELL Wnt Pathway library v 2.0 is a focused collection of 71 compounds with defined and diverse Wnt pathway activity, including activators and inhibitors of Wnts, Dishevelled, GSK-3β and other Wnt pathway kinases, TCF/β-catenin, DKK, LRP, Axin, Porcupine, and more. Compounds are dissolved in DMSO at 10mM or 1mM and aliquoted into deep-well plates at 100 or 500µl per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-Wnt pathway molecules in cells as well as for use in in vitro applications. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0216 Supplier Enzo Life Sciences Supplier No. BML28380500
The SCREEN-WELL Wnt Pathway library v 2.0 is a focused collection of 71 compounds with defined and diverse Wnt pathway activity, including activators and inhibitors of Wnts, Dishevelled, GSK-3β and other Wnt pathway kinases, TCF/β-catenin, DKK, LRP, Axin, Porcupine, and more. Compounds are dissolved in DMSO at 10mM or 1mM and aliquoted into deep-well plates at 100 or 500µl per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-Wnt pathway molecules in cells as well as for use in in vitro applications. Long Term Storage: -80°C. Applications: HTS .