Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Wnt Surrogate-Fc Fusion Recombinant Protein

Catalog No. PHG0403
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHG0403 500 μg
PHG0401 25 μg
PHG0402 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PHG0403 Supplier Gibco™ Supplier No. PHG0403
  • $4,030.00
    $3,828.65 / Each of 1

    Save $201.35

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Gibco Wnt Surrogate-Fc Fusion Protein activates similar Wnt signaling pathways as most commercially available recombinant Wnt3a proteins or Wnt-conditioned media, while requiring a fraction of the amount of protein or hassle. Gibco Wnt Surrogate-Fc Fusion Protein was engineered with improved solubility, overcoming a major obstacle of working with Wnt3a and reducing the concentrations required to activate Wnt signaling in your model system. Gibco Wnt Surrogate-Fc Fusion Protein is comprised of the LRP-binding domain of DKK linked to a selective, high affinity binder of Frizzled receptors. Gibco Wnt Surrogate-Fc Fusion Protein is provided at a concentration of 2 mg/mL which is equivalent to 17.1 μM based on dimeric stoichiometry. Storage: Store undiluted Gibco Wnt Surrogate-Fc Fusion Protein at -80° for up to 1 year. After thawing, apportion into working aliquots and store at -20°C to -80°C for up to 2 months.

Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P56704
Concentration 2 mg/mL
For Use With (Application) Bioactivity
Formulation 25mM tris with 500mM NaCl and no preservative; pH 8.2
Gene ID (Entrez) 89780
Name Wnt Surrogate-Fc Fusion
pH Range 8.2
Purification Method Purified
Quantity 500 μg
Source HEK293 cells
Storage Requirements -20°C or -80°C if preferred
Regulatory Status RUO
Endotoxin Concentration 1 EU/mL
Gene Alias MGC119418; MGC119419; MGC119420; Protein Wnt-3a; vestigial tail; vt; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3A; Wnt 3a; Wnt family member 3A; wnt[a]; wnt3; wnt3 l; wnt3 like; Wnt3a; Wnt-3a; WNT3A protein; wnt3l; Zwnt[a]
Common Name WNT3A
Gene Symbol WNT3A
Biological Activity Greater than 25 fold induction in Topflash assay activity (at a Wnt surrogate-Fc protein concentration between 0.03 - 5.0 nM (based on dimeric stoichiometry))
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Content And Storage -20°Cor to +80°C if preferred
Activity Greater than 25 fold induction in Topflash assay activity at a Wnt surrogate-Fc protein concentration between 0.03 - 5.0 nM (based on dimeric stoichiometry)
Expression System HEK293 cells
Form Liquid
Purity or Quality Grade > 95%
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.