Learn More
Aviva Systems Biology Corp WNT2B Antibody - middle region (ARP41254_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41254_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against WNT2B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human WNT2B. Alternate names: WNT13, XWNT2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53291