Learn More
Aviva Systems Biology Corp WNT2B Antibody - N-terminal region (ARP41275_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41275_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WNT2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WNT2B. Alternate names: WNT13, XWNT2ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Human, Rabbit
Catalog No. 50-014-57789