Learn More
Aviva Systems Biology Corp WNT3 Antibody - middle region (ARP49956_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49956_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WNT3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human WNT3. Alternate names: INT4, MGC131950, MGC138321, MGC138323ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-468