Learn More
Aviva Systems Biology Corp Wnt3a Antibody - C-terminal region (ARP58819_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58819_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Wnt3a. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: vt, Wnt-3aApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-499