Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Wnt3a Surrogate Hum 100ug
SDP

Catalog No. 5000184666
Encompass_Preferred

Recombinant human Wnt3a surrogate Fc-fusion protein expressed in HEK293 cells, supplied as a filtered, buffered solution for research use in Wnt signaling and related cell biology assays. Supplied in a 100 μg format and intended for applications requiring high-purity, low-endotoxin protein.

  • Human Fc fusion expressed in HEK293 cells.
  • Available as a 100 μg vial for assay use.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin below 0.01 EU/μg (LAL test).
  • Formulated in 25 mM Tris-HCl, 500 mM NaCl, pH 8.2, 0.2 μm filtered.
  • Predicted molecular mass 58.5 kDa; apparent 66-73 kDa on SDS-PAGE.
  • Store at -80°C for long-term storage; avoid repeated freeze-thaw cycles.

Catalog No. 50-001-84666 Supplier Medchemexpress LLC Supplier No. HYP70453B100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Wnt3a surrogate Fc-fusion protein expressed in HEK293 cells, supplied as a filtered, buffered solution for research use in Wnt signaling and related cell biology assays. Supplied in a 100 μg format and intended for applications requiring high-purity, low-endotoxin protein.

  • Human Fc fusion expressed in HEK293 cells.
  • Available as a 100 μg vial for assay use.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin below 0.01 EU/μg (LAL test).
  • Formulated in 25 mM Tris-HCl, 500 mM NaCl, pH 8.2, 0.2 μm filtered.
  • Predicted molecular mass 58.5 kDa; apparent 66-73 kDa on SDS-PAGE.
  • Store at -80°C for long-term storage; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.