Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Wnt3a Surrogate Hum 50ug
SDP

Catalog No. 5000184959
Encompass_Preferred

Wnt3a Surrogate Protein, Human (HEK293, C-hFc) is a recombinant, Fc-tagged surrogate of human Wnt3a produced in HEK293 cells. Supplied lyophilized, it serves as a water-soluble, Frizzled-specific Wnt agonist for in vitro research to activate Wnt signaling pathways.

  • Expressed in HEK293 cells with a C-terminal human Fc tag.
  • Supplied lyophilized from PBS (pH 7.4) with 8% trehalose.
  • Purity greater than 95% by Tris-Bis PAGE.
  • Predicted molecular mass 58.5 kDa; appears ~60-75 kDa on SDS-PAGE.
  • Recommended reconstitution concentration of at least 100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • For research use only.

Catalog No. 50-001-84959 Supplier Medchemexpress LLC Supplier No. HYP70453C50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wnt3a Surrogate Protein, Human (HEK293, C-hFc) is a recombinant, Fc-tagged surrogate of human Wnt3a produced in HEK293 cells. Supplied lyophilized, it serves as a water-soluble, Frizzled-specific Wnt agonist for in vitro research to activate Wnt signaling pathways.

  • Expressed in HEK293 cells with a C-terminal human Fc tag.
  • Supplied lyophilized from PBS (pH 7.4) with 8% trehalose.
  • Purity greater than 95% by Tris-Bis PAGE.
  • Predicted molecular mass 58.5 kDa; appears ~60-75 kDa on SDS-PAGE.
  • Recommended reconstitution concentration of at least 100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.