Learn More
Aviva Systems Biology Corp WNT5A Antibody - middle region (ARP32128_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32128_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WNT5A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human WNT5A. Alternate names: hWNT5AApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52854