Learn More
Aviva Systems Biology Corp WNT5B Antibody - C-terminal region (ARP41280_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41280_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against WNT5B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human WNT5B. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57792