Learn More
Medchemexpress LLC Wnt6 protein, mouse (His, SUMO) | >90.0% | 100UG

Supplier: Medchemexpress LLC HYP702628100UG
Wnt6 Protein, Mouse (His, SUMO) is a recombinant mouse-derived Wnt6 produced in Escherichia coli and supplied lyophilized. The protein contains N-terminal SUMO and 6xHis tags, has a predicted molecular mass of 50.2 kDa (≈52 kDa by SDS-PAGE), and is reported to be >90% pure by reducing SDS-PAGE. It is supplied with trehalose-containing buffer excipients and is intended for research use; follow the supplier's reconstitution guidance (do not reconstitute below 100 μg/mL).
- Recombinant mouse Wnt6 expressed in E. coli.
- N-terminal SUMO and 6xHis affinity tags.
- Predicted molecular weight ~50.2 kDa (≈52 kDa by SDS-PAGE).
- Purity greater than 90% by reducing SDS-PAGE.
- Lyophilized with trehalose-containing storage buffer for stability.
- Available in common research sizes (20 μg, 50 μg, 100 μg).
- For research use only; follow reconstitution and handling instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.