Learn More
Aviva Systems Biology Corp WNT9B Antibody - C-terminal region (ARP41243_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41243_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against WNT9B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human WNT9B. Alternate names: WNT15, WNT14BApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-57777