Learn More
Aviva Systems Biology Corp WNT9B Antibody - middle region (ARP41242_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41242_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WNT9B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human WNT9B. Alternate names: WNT14B, WNT15ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57776