Learn More
Sino Biological WNV WNV-E MAb 100ul (Sino Biological)
Supplier:  Sino Biological 40345MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant West Nile Virus (WNV) (lineage 1, strain NY99) E / Envelope (Domain III) ( Catalog#40345-V08Y1; ABA62343.1; Gln586-Ala705). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3156