Learn More
Research Products International Corp Wobble-Not SERO Pipets, Shorty, 10ml, 200/pk
SDP
Catalog No. 502229463
Encompass
Wobble-Not Serological Pipets feature innovative plug-end design that require less force when inserting to pipet controller. Pipets offer an excellent secure seal, maximum stability, better accuracy and ultra-clear markings. Supplied individually wrapped (paper/plastic) with porous polypropylene filter and ±2% accuracy at full volume.
Snug, tight fit
Drip free
RNase/DNase free
Sterile, non-pyrogenic
Catalog No. 50-222-9463 Supplier Research Products International Corp Supplier No. 163258
Wobble-Not Serological Pipets feature innovative plug-end design that require less force when inserting to pipet controller. Pipets offer an excellent secure seal, maximum stability, better accuracy and ultra-clear markings. Supplied individually wrapped (paper/plastic) with porous polypropylene filter and ±2% accuracy at full volume.
Snug, tight fit
Drip free
RNase/DNase free
Sterile, non-pyrogenic