Learn More
Selleck Chemical LLC Wogonin-10mg
SDP
Supplier:  Selleck Chemical LLC S474310MG
Encompass_Preferred
Wogonin (Vogonin), a natural and biologically-active flavonoid found in plants, is an inhibitor of CDK9 and does not inhibit CDK2, CDK4 and CDK6 at doses that inhibit CDK9 activity; Also inhibits N-acetyltransferase.
Catalog No. 50-238-9817