Learn More
AdipoGen Wogonin (25 mg)
SDP
Supplier:  AdipoGen AGCN20064M025
Encompass_Preferred
Chemical. CAS: 632-85-9. Formula: C16H12O5. MW: 284.3. Isolated from Scutellaria baicalensis Georgi. Anti-inflammatory. Increases nitric oxide (NO) production. Inhibits PGE2, cyclooxygenase-2 (COX-2) and inducible nitric oxide synthase (iNOS; NOS II). Antioxidant. Anti-viral. Shows anti-hepatitis B virus activity. MCP-1 inhibitor. Shows anxiolytic effect through modulation of the GABA(A) receptor complex. Antifungal. Neuroprotective. Anticancer compound. Apoptosis and cell cycle arrest inducer. Sensitizes TNFalpha- and TRAIL-induced apoptosis. Antiangiogenic. NF-kappaB inhibitor. Telomerase activity inhibitor. CDK9 inhibitor. PI3K-AKT pathway modulator. Autophagy inducer.
Catalog No. 50-205-3411