Learn More
MSC Wontide Telescoping Lift Master Boom, 6000 lb, 80-144 in Length
Catalog No. 5000514407
Encompass
The telescoping lift master boom converts fork trucks into overhead lifting cranes. With a 6000 lb capacity and an adjustable telescoping length of 80-144 inches, this durable boom provides a robust solution for various material handling needs.
  • Converts fork trucks into overhead lifting cranes.
  • Fork pockets with safety strap secure the boom to the fork truck.
  • Robotically welded construction ensures precision and durability.
  • Operations and capacity sticker provide user instructions and safety.
  • Fabricated from structural steel with ISO compliant welding for long-term service.
  • Infinitely adjustable locking screw on telescopic units.
Catalog No. 50-005-14407 Supplier MSC Supplier No. 47345814
The telescoping lift master boom converts fork trucks into overhead lifting cranes. With a 6000 lb capacity and an adjustable telescoping length of 80-144 inches, this durable boom provides a robust solution for various material handling needs.
  • Converts fork trucks into overhead lifting cranes.
  • Fork pockets with safety strap secure the boom to the fork truck.
  • Robotically welded construction ensures precision and durability.
  • Operations and capacity sticker provide user instructions and safety.
  • Fabricated from structural steel with ISO compliant welding for long-term service.
  • Infinitely adjustable locking screw on telescopic units.