Learn More
Andwin Scientific WOOD APPLICATOR
SDP
Catalog No. NC9980692
Encompass_Preferred
The disposable wooden Puritan Applicators feature a uniform cylindrical shape and smooth finish. This product is ideal for routine spot test sampling and drop transfers, or wherever a handy, disposable mixing aid is required. The applicators are dust-free and are measured to have the dimensions of 147.6 x 2.1 mm (513/16 x 3/32").
This item is not returnable. View return policy
Catalog No. NC9980692 Supplier Andwin Scientific Supplier No. 10805018
The disposable wooden Puritan Applicators feature a uniform cylindrical shape and smooth finish. This product is ideal for routine spot test sampling and drop transfers, or wherever a handy, disposable mixing aid is required. The applicators are dust-free and are measured to have the dimensions of 147.6 x 2.1 mm (513/16 x 3/32").