Learn More
Electron Microscopy Sciences Wood Applicators, 12"L
SDP
Catalog No. 50949238
Encompass_Preferred
These sticks are made from closed-grain Northern White Birch. This selective wood provides a smooth, splinter-free finish with good tensile strength. In addition to medical uses, these sticks are ideal for spreading ointments and salves, stirring and mixing and general laboratory applications.
OAL: 7"
Diameter: 5/32"
Catalog No. 50-949-238 Supplier Electron Microscopy Sciences Supplier No. 72303
These sticks are made from closed-grain Northern White Birch. This selective wood provides a smooth, splinter-free finish with good tensile strength. In addition to medical uses, these sticks are ideal for spreading ointments and salves, stirring and mixing and general laboratory applications.
OAL: 7"
Diameter: 5/32"