Learn More
Electron Microscopy Sciences Wood Applicators, 6"L
SDP
Catalog No. 501899814
Encompass_Preferred
These sticks are made from closed-grain Northern White Birch. This selective wood provides a smooth, splinter-free finish with good tensile strength. In addition to medical uses, these sticks are ideal for spreading ointments and salves, stirring and mixing and general laboratory applications.
OAL: 6"
Diameter: 1/2"
Catalog No. 50-189-9814 Supplier Electron Microscopy Sciences Supplier No. 72300
These sticks are made from closed-grain Northern White Birch. This selective wood provides a smooth, splinter-free finish with good tensile strength. In addition to medical uses, these sticks are ideal for spreading ointments and salves, stirring and mixing and general laboratory applications.
OAL: 6"
Diameter: 1/2"