Learn More
Andwin Scientific WOOD TONGUE DEPRESSOR 1000/PK
SDP
Catalog No. 50949206
Encompass_Preferred
The wooden sticks are ideal for use as tongue depressors. Constructed with close-grained northern white birch, the wooden sticks feature a smooth and polished surface. This product can also be utilized for mixing, stirring, or scraping epoxy resins. It has a dimension of 4.5" x 3/8". It is sold in a pack of 1000.
This item is not returnable. View return policy
Catalog No. 50-949-206 Supplier Andwin Scientific Supplier No. 72322
The wooden sticks are ideal for use as tongue depressors. Constructed with close-grained northern white birch, the wooden sticks feature a smooth and polished surface. This product can also be utilized for mixing, stirring, or scraping epoxy resins. It has a dimension of 4.5" x 3/8". It is sold in a pack of 1000.