Learn More
Diversified Spaces™ Wood Workbench with Maple Top
SCI_ED
Catalog No. S42630 Shop All Diversified Spaces Products
Change view
Click to view available options
Dimensions (D x W x H):
30 x 72 x 30.75 in.
36 x 60 x 30.75 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
S42630 36 x 60 x 30.75 in.
S42631 30 x 72 x 30.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S42630 Supplier Diversified Spaces™ Supplier No. W6036LM
Add to Cart
Add to Cart
Workbench with maple plywood shelves, solid maple legs and stringers available in customizable lengths.
  • Solid maple legs and stringers with maple plywood shelf
  • Constructed from solid maple and maple veneers
  • Durable earth-friendly UV finish
  • SEFA compliant and MAS Certified Green
Specifications
Product Type Table
Color Maple
Dimensions (D x W x H) 36 x 60 x 30.75 in.
Shelf Material Maple Plywood
Warranty Limited Lifetime
Material Maple, Maple Veneer