Learn More
Electron Microscopy Sciences Wood Applicator Stick, 6" x 3/32"
SDP
Catalog No. NC1563779
Encompass_Preferred
These sticks are made from closed-grain Northern White Birch. This selective wood provides a smooth, splinter-free finish with good tensile strength. In addition to medical uses, these sticks are ideal for spreading ointments and salves, stirring and mixing and general laboratory applications.
OAL: 6"
Diameter: 3/32"
Catalog No. NC1563779 Supplier Electron Microscopy Sciences Supplier No. 72300SPF
These sticks are made from closed-grain Northern White Birch. This selective wood provides a smooth, splinter-free finish with good tensile strength. In addition to medical uses, these sticks are ideal for spreading ointments and salves, stirring and mixing and general laboratory applications.
OAL: 6"
Diameter: 3/32"