Learn More
Electron Microscopy Sciences Wooden Picks, Round, Double Tapered, 2pk/100
SDP
Catalog No. 50949075
Encompass_Preferred
They are round and double tapered; used for cleaning pole pieces and parts of the EM. Also used for orienting the specimen during the embedding stage.
Catalog No. 50-949-075 Supplier Electron Microscopy Sciences Supplier No. 72340
They are round and double tapered; used for cleaning pole pieces and parts of the EM. Also used for orienting the specimen during the embedding stage.