Learn More
Electron Microscopy Sciences Wooden Tweezers Holder Case
SDP
Catalog No. 5024049
Encompass_Preferred
Made from solid wood, with brass double hinges and an elegant latch. Comes with a foam padded top and bottom to protect stored tweezers. The inside measures 5 3/4" x 4 7/8" (145 x 124 mm) and it holds 5 tweezers with a maximum length of 140 mm.
Catalog No. 50-240-49 Supplier Electron Microscopy Sciences Supplier No. 7289801
Made from solid wood, with brass double hinges and an elegant latch. Comes with a foam padded top and bottom to protect stored tweezers. The inside measures 5 3/4" x 4 7/8" (145 x 124 mm) and it holds 5 tweezers with a maximum length of 140 mm.