Learn More
CPC Scientific 5FAM-Lys-Lys-Ile-Ser-Gly-Arg-Leu-Ser-Pro-Ile-Met-Thr-Glu-Gln-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110498
Encompass
FAM-labeled Forkhead derived peptide (Ab/Em=494/521 nm) also known as woodtide. This peptide is used as a substrate for the DYRK family of kinases in in-vitro analysis.ONE-LETTER SEQUENCE: 5FAM-KKISGRLSPIMTEQ-NH2MOLECULAR FORMULA: C89H133N21O26S1MOLECULAR WEIGHT:1945.20STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1566528-51-5], SYNONYMS: 5-FAM-Woodtide trifluoroacetate salt Fluorescein-5-carbonyl-Lys-Lys-Ile-Ser-Gly-Arg-Leu-Ser-Pro-Ile-Met-Thr-Glu-Gln-NH2RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Woods, Y. et al. Biochem J. 355, 597(2001); Woods, Y. et al. Biochem. J. 355, 609(2001); Klumpp, M. et al. J. Biomol. Screen. 11, 617(2006)
Catalog No. 50-211-0498 Supplier CPC Scientific Supplier No. SUBS020A
FAM-labeled Forkhead derived peptide (Ab/Em=494/521 nm) also known as woodtide. This peptide is used as a substrate for the DYRK family of kinases in in-vitro analysis.ONE-LETTER SEQUENCE: 5FAM-KKISGRLSPIMTEQ-NH2MOLECULAR FORMULA: C89H133N21O26S1MOLECULAR WEIGHT:1945.20STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1566528-51-5], SYNONYMS: 5-FAM-Woodtide trifluoroacetate salt Fluorescein-5-carbonyl-Lys-Lys-Ile-Ser-Gly-Arg-Leu-Ser-Pro-Ile-Met-Thr-Glu-Gln-NH2RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Woods, Y. et al. Biochem J. 355, 597(2001); Woods, Y. et al. Biochem. J. 355, 609(2001); Klumpp, M. et al. J. Biomol. Screen. 11, 617(2006)