Learn More
MSC Bally Block Workbench Top, Maple, 1-3/4 in. x 30 in. x 60 in., 1 ea.
Catalog No. 5000516747
Encompass
This durable maple workbench top provides a robust and reliable work surface for industrial and laboratory workstations. Constructed as a single, warp-resistant, self-supporting panel, this top is designed for longevity and heavy-duty use in demanding environments.
  • Features a thickness of 1-3/4 in.
  • Dimensions measure 30 in. x 60 in.
  • Finished with an ultraviolet cured acrylic coating for enhanced durability
  • Resistant to water, alcohol, and solvent detergents
  • Machined and sanded to a smooth, satin finish for immediate installation
  • Made in the USA
Catalog No. 50-005-16747 Supplier MSC Supplier No. 09783911
This durable maple workbench top provides a robust and reliable work surface for industrial and laboratory workstations. Constructed as a single, warp-resistant, self-supporting panel, this top is designed for longevity and heavy-duty use in demanding environments.
  • Features a thickness of 1-3/4 in.
  • Dimensions measure 30 in. x 60 in.
  • Finished with an ultraviolet cured acrylic coating for enhanced durability
  • Resistant to water, alcohol, and solvent detergents
  • Machined and sanded to a smooth, satin finish for immediate installation
  • Made in the USA