Learn More
Grainger Little Giant Work Height Platform Truck, 2,000 lb, Steel, 60 in x 30 in, 1/Each
Catalog No. 502784438
Encompass_Preferred
This heavy-duty steel-deck platform truck is designed for ergonomic efficiency, featuring an adjustable deck height that minimizes lifting and bending when transferring items between the truck and elevated work surfaces. The robust 12 ga solid steel deck is powder-coated for durability and smooth operation in industrial environments.
  • Load capacity of 2,000 lb
  • Adjustable deck height ranging from 24 in. to 34 in.
  • Includes four 6 in. mold-on rubber casters in a configuration of two rigid and two swivel
  • Removable raised handle located at a single end
  • Solid flush deck surface with powder-coated finish
  • Assembled construction
Catalog No. 50-278-4438 Supplier Grainger Supplier No. 19G899
This heavy-duty steel-deck platform truck is designed for ergonomic efficiency, featuring an adjustable deck height that minimizes lifting and bending when transferring items between the truck and elevated work surfaces. The robust 12 ga solid steel deck is powder-coated for durability and smooth operation in industrial environments.
  • Load capacity of 2,000 lb
  • Adjustable deck height ranging from 24 in. to 34 in.
  • Includes four 6 in. mold-on rubber casters in a configuration of two rigid and two swivel
  • Removable raised handle located at a single end
  • Solid flush deck surface with powder-coated finish
  • Assembled construction