Learn More
Grainger Workbench Butcher Block 60 W 30 D
Catalog No. 502768115
Encompass_Preferred
The Pro-Line Workbench with Adjustable Height is a heavy-duty steel workbench that provides a durable and stable work surface. It features a 1-1/4" thick maple laminate top that measures 60" W x 24" D, providing ample workspace. The height is adjustable from 30-1/2" to 37-1/2" using a foot pedal to accommodate sitting or standing. The steel frame with powder coat finish provides 800 lb weight capacity. It's an ideal industrial workbench for garage, shop, or warehouse use.
Catalog No. 50-276-8115 Supplier Grainger Supplier No. 16Y947
Add to Cart
Add to Cart
Request a Quote
The Pro-Line Workbench with Adjustable Height is a heavy-duty steel workbench that provides a durable and stable work surface. It features a 1-1/4" thick maple laminate top that measures 60" W x 24" D, providing ample workspace. The height is adjustable from 30-1/2" to 37-1/2" using a foot pedal to accommodate sitting or standing. The steel frame with powder coat finish provides 800 lb weight capacity. It's an ideal industrial workbench for garage, shop, or warehouse use.