Learn More
Grainger Workbench ESD Laminate 120 W 36 D
Catalog No. 502766602
Encompass_Preferred
The BENCHPRO Workbench with Adjustable Height is a sturdy steel workbench that allows you to adjust the height from 30" to 44" to accommodate sitting or standing. It has a 1-1/4" thick maple laminate worksurface that measures 60" W x 24" D, providing ample workspace. The bench also features four heavy-duty swivel casters with locks so you can easily move it around your workspace. This high-quality workbench is made in the USA and can support up to 1000 lbs, making it ideal for garage, shop, or industrial use.
Catalog No. 50-276-6602 Supplier Grainger Supplier No. 16Y096
The BENCHPRO Workbench with Adjustable Height is a sturdy steel workbench that allows you to adjust the height from 30" to 44" to accommodate sitting or standing. It has a 1-1/4" thick maple laminate worksurface that measures 60" W x 24" D, providing ample workspace. The bench also features four heavy-duty swivel casters with locks so you can easily move it around your workspace. This high-quality workbench is made in the USA and can support up to 1000 lbs, making it ideal for garage, shop, or industrial use.