Learn More
Grainger Workbench ESD Laminate 60 L x 24 D
Catalog No. 502768005
Encompass_Preferred
The Grainger 16Y089 configurable workbench leg rests and height extenders are accessories compatible with Grainger's line of modular workbenches. They allow you to raise the height of the work surface from 30" up to 40" in 2" increments. The leg rests also provide a foot rest bar in front of the bench. They are made from heavy gauge steel with a powder coat finish and install easily by attaching to the underside of the workbench. These accessories add comfort and ergonomics by allowing you to customize the height of your workbench.
Catalog No. 50-276-8005 Supplier Grainger Supplier No. 16Y089
The Grainger 16Y089 configurable workbench leg rests and height extenders are accessories compatible with Grainger's line of modular workbenches. They allow you to raise the height of the work surface from 30" up to 40" in 2" increments. The leg rests also provide a foot rest bar in front of the bench. They are made from heavy gauge steel with a powder coat finish and install easily by attaching to the underside of the workbench. These accessories add comfort and ergonomics by allowing you to customize the height of your workbench.