Learn More
Grainger Workbench ESD Laminate 72 W 30 D
Catalog No. 502767814
Encompass_Preferred
The BENCHPRO Workbench with Adjustable Height is a sturdy steel workbench that allows you to adjust the height from 30" to 42" to accommodate sitting or standing. It has a 1-1/4" thick maple laminate work surface that measures 60" W x 24" D and can hold up to 1200 lbs. The bench also features four heavy-duty swivel casters with locks for easy mobility. This industrial-grade workbench is ideal for garage, shop, or warehouse use.
Catalog No. 50-276-7814 Supplier Grainger Supplier No. 16Y091
The BENCHPRO Workbench with Adjustable Height is a sturdy steel workbench that allows you to adjust the height from 30" to 42" to accommodate sitting or standing. It has a 1-1/4" thick maple laminate work surface that measures 60" W x 24" D and can hold up to 1200 lbs. The bench also features four heavy-duty swivel casters with locks for easy mobility. This industrial-grade workbench is ideal for garage, shop, or warehouse use.