Learn More
Grainger Workbench ESD Laminate 72 W 36 D
Catalog No. 502766597
Encompass_Preferred
The Pro-Line L-Shaped Workbench from Grainger is a fixed-height workbench made from steel with a powder-coated finish. It features an L-shaped design with two work surfaces that measure 60"W x 24"D and 48"W x 24"D. The total dimensions are 60"W x 48"D x 34"H. This industrial-grade workbench can support up to 2500 lbs and comes equipped with (2) shelves, (6) drawers, and (6) pegboard panels for storage and organization. The workbench requires assembly.
Catalog No. 50-276-6597 Supplier Grainger Supplier No. 16Y094
The Pro-Line L-Shaped Workbench from Grainger is a fixed-height workbench made from steel with a powder-coated finish. It features an L-shaped design with two work surfaces that measure 60"W x 24"D and 48"W x 24"D. The total dimensions are 60"W x 48"D x 34"H. This industrial-grade workbench can support up to 2500 lbs and comes equipped with (2) shelves, (6) drawers, and (6) pegboard panels for storage and organization. The workbench requires assembly.