Learn More
Grainger Workbench ESD Laminate 96 W 30 D
Catalog No. 502766596
Encompass_Preferred
The BENCHPRO Workbench with Adjustable Height is a sturdy steel workbench that allows you to adjust the height from 30" to 44" to accommodate sitting or standing. It has a 1-1/4" thick maple laminate worksurface that measures 60"W x 24"D. The steel frame with powder coat finish provides 800 lb. load capacity. It also features four heavy-duty swivel casters with locks for easy mobility. This height adjustable workbench is ideal for industrial, technical and engineering workspaces.
Catalog No. 50-276-6596 Supplier Grainger Supplier No. 16Y092
The BENCHPRO Workbench with Adjustable Height is a sturdy steel workbench that allows you to adjust the height from 30" to 44" to accommodate sitting or standing. It has a 1-1/4" thick maple laminate worksurface that measures 60"W x 24"D. The steel frame with powder coat finish provides 800 lb. load capacity. It also features four heavy-duty swivel casters with locks for easy mobility. This height adjustable workbench is ideal for industrial, technical and engineering workspaces.