Learn More
Grainger Workbench Laminate 60 W 30 D
Catalog No. 502766928
Encompass_Preferred
The Workbench Top Laminate from Grainger is a durable and easy to clean laminate work surface ideal for garage, shop, or industrial workbenches. It measures 96" long x 25" wide x 1-1/4" thick and features a smooth, black laminate finish over industrial grade particleboard. The 1,250 lb weight capacity ensures stability for heavy-duty tasks. Pre-drilled holes allow secure mounting to a workbench frame. The scratch and scuff resistant laminate cleans easily. This workbench top provides an affordable and functional work surface.
Catalog No. 50-276-6928 Supplier Grainger Supplier No. 16Y074
The Workbench Top Laminate from Grainger is a durable and easy to clean laminate work surface ideal for garage, shop, or industrial workbenches. It measures 96" long x 25" wide x 1-1/4" thick and features a smooth, black laminate finish over industrial grade particleboard. The 1,250 lb weight capacity ensures stability for heavy-duty tasks. Pre-drilled holes allow secure mounting to a workbench frame. The scratch and scuff resistant laminate cleans easily. This workbench top provides an affordable and functional work surface.