Learn More
Grainger Workbench Laminate 60 W 30 D
Catalog No. 502767936
Encompass_Preferred
The Pro-Line L-Shaped Workbench from Grainger is a fixed-height workbench made from steel with a powder-coated finish. It features an L-shaped design with two work surfaces that measure 60"W x 24"D and 48"W x 24"D. The total dimensions are 60"W x 48"D x 34"H. This industrial-grade workbench can support up to 1200 lbs and comes equipped with (2) lower shelves for storage. The work surface is scratch resistant and the bench has leveling feet for stability on uneven floors. This is a high-quality, durable workbench well-suited for garage, shop, or industrial use.
Catalog No. 50-276-7936 Supplier Grainger Supplier No. 16Y946
Add to Cart
Add to Cart
Request a Quote
The Pro-Line L-Shaped Workbench from Grainger is a fixed-height workbench made from steel with a powder-coated finish. It features an L-shaped design with two work surfaces that measure 60"W x 24"D and 48"W x 24"D. The total dimensions are 60"W x 48"D x 34"H. This industrial-grade workbench can support up to 1200 lbs and comes equipped with (2) lower shelves for storage. The work surface is scratch resistant and the bench has leveling feet for stability on uneven floors. This is a high-quality, durable workbench well-suited for garage, shop, or industrial use.