Learn More
Grainger Workbench Laminate 60 W 30 D
Catalog No. 502767938
Encompass_Preferred
The Pro-Line Workbench Fixed Height is a heavy-duty steel workbench that provides a durable and stable work surface. It features an all-welded steel frame with leveling feet, and a 1-1/4" thick maple laminate work surface that is oil, scratch, and impact resistant. The workbench has a fixed 30" working height, and is available in lengths of 4', 5', 6', and 8', with depths of 24" or 30". Other details include a lower steel shelf for storage, and pre-drilled holes for adding a vise. This rugged workbench is ideal for garage, shop, or industrial use.
Catalog No. 50-276-7938 Supplier Grainger Supplier No. 16Y913
The Pro-Line Workbench Fixed Height is a heavy-duty steel workbench that provides a durable and stable work surface. It features an all-welded steel frame with leveling feet, and a 1-1/4" thick maple laminate work surface that is oil, scratch, and impact resistant. The workbench has a fixed 30" working height, and is available in lengths of 4', 5', 6', and 8', with depths of 24" or 30". Other details include a lower steel shelf for storage, and pre-drilled holes for adding a vise. This rugged workbench is ideal for garage, shop, or industrial use.