Learn More
Grainger Workbench Laminate 60 W 30 D
Catalog No. 502767606
Encompass_Preferred
The Pro-Line Workbench Fixed Height is a sturdy steel workbench that provides a durable and stable work surface. It features an all-welded steel frame with leveling feet and measures 60"W x 24"D x 34"H. The 1-1/4" thick maple laminate worksurface offers chemical, impact, and scratch resistance. This industrial-grade workbench can support up to 2500 lbs and comes with pre-drilled holes to mount vises, shelves, and other accessories. Its powder coated finish resists chipping, scratching, and corrosion. This is an ideal heavy-duty workbench for garage, shop, or industrial use.
Catalog No. 50-276-7606 Supplier Grainger Supplier No. 16Y911
The Pro-Line Workbench Fixed Height is a sturdy steel workbench that provides a durable and stable work surface. It features an all-welded steel frame with leveling feet and measures 60"W x 24"D x 34"H. The 1-1/4" thick maple laminate worksurface offers chemical, impact, and scratch resistance. This industrial-grade workbench can support up to 2500 lbs and comes with pre-drilled holes to mount vises, shelves, and other accessories. Its powder coated finish resists chipping, scratching, and corrosion. This is an ideal heavy-duty workbench for garage, shop, or industrial use.