Learn More
Grainger Workbench Laminate 60 W 36 D
Catalog No. 502767608
Encompass_Preferred
The Pro-Line Workbench Fixed Height is a sturdy steel workbench that provides a durable and stable work surface. It has a 1-1/4" thick maple laminate worktop that measures 60"W x 24"D. The bench height is 34" and cannot be adjusted. The steel frame and legs have a powder coated finish to prevent rust and corrosion. It can support up to 1200 lbs of weight evenly distributed. There are 2 lower shelves for extra storage space. Assembly is required upon delivery. This industrial grade workbench is ideal for garage, shop, or warehouse use.
Catalog No. 50-276-7608 Supplier Grainger Supplier No. 16Y915
The Pro-Line Workbench Fixed Height is a sturdy steel workbench that provides a durable and stable work surface. It has a 1-1/4" thick maple laminate worktop that measures 60"W x 24"D. The bench height is 34" and cannot be adjusted. The steel frame and legs have a powder coated finish to prevent rust and corrosion. It can support up to 1200 lbs of weight evenly distributed. There are 2 lower shelves for extra storage space. Assembly is required upon delivery. This industrial grade workbench is ideal for garage, shop, or warehouse use.