Learn More
Grainger Workbench Laminate 72 W 30 D
Catalog No. 502767607
Encompass_Preferred
The Pro-Line L-Shaped Workbench from Grainger is a fixed-height workbench made of heavy-duty steel with a powder-coated finish. It features an L-shaped design with two work surfaces that measure 60"W x 24"D and 48"W x 24"D. The total dimensions are 60"W x 48"D x 34"H. This industrial-grade workbench can support up to 2500 lbs and comes equipped with (2) shelves, (6) drawers, and multiple pegboard storage options for organizing tools and supplies. Its sturdy steel construction and versatile storage make it well-suited for garage, shop, or industrial use.
Catalog No. 50-276-7607 Supplier Grainger Supplier No. 16Y914
The Pro-Line L-Shaped Workbench from Grainger is a fixed-height workbench made of heavy-duty steel with a powder-coated finish. It features an L-shaped design with two work surfaces that measure 60"W x 24"D and 48"W x 24"D. The total dimensions are 60"W x 48"D x 34"H. This industrial-grade workbench can support up to 2500 lbs and comes equipped with (2) shelves, (6) drawers, and multiple pegboard storage options for organizing tools and supplies. Its sturdy steel construction and versatile storage make it well-suited for garage, shop, or industrial use.