Learn More
Grainger Workbench Laminate 72 W 30 D
Catalog No. 502767438
Encompass_Preferred
The Pro-Line Workbench Fixed Height is a heavy-duty steel workbench that provides a durable and stable work surface. It features an all-welded steel frame with leveling feet and measures 60"W x 24"D x 34"H. The 1-1/4" thick maple laminate work surface provides ample workspace. This industrial-grade workbench can support up to 2500 lbs and is designed for garage, shop, and warehouse use. With its fixed height design, this sturdy workbench creates an ideal flat workspace for various projects and tasks.
Catalog No. 50-276-7438 Supplier Grainger Supplier No. 16Y912
The Pro-Line Workbench Fixed Height is a heavy-duty steel workbench that provides a durable and stable work surface. It features an all-welded steel frame with leveling feet and measures 60"W x 24"D x 34"H. The 1-1/4" thick maple laminate work surface provides ample workspace. This industrial-grade workbench can support up to 2500 lbs and is designed for garage, shop, and warehouse use. With its fixed height design, this sturdy workbench creates an ideal flat workspace for various projects and tasks.