Learn More
Grainger Workbench Laminate 96 W 30 D
Catalog No. 502766593
Encompass_Preferred
The Grainger 16Y076 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for a variety of cleaning applications. This 8" x 4" hand pad has an open web construction that allows it to conform to curved surfaces. It is made of high-quality fibers that are durable and long-lasting. The pad is designed for scrubbing and cleaning while being gentle on surfaces. It can be used wet or dry on a variety of materials including stainless steel, aluminum, ceramic, plastic, glass and painted wood. This hand pad from 3M and Grainger provides an effective cleaning solution.
Catalog No. 50-276-6593 Supplier Grainger Supplier No. 16Y076
The Grainger 16Y076 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for a variety of cleaning applications. This 8" x 4" hand pad has an open web construction that allows it to conform to curved surfaces. It is made of high-quality fibers that are durable and long-lasting. The pad is designed for scrubbing and cleaning while being gentle on surfaces. It can be used wet or dry on a variety of materials including stainless steel, aluminum, ceramic, plastic, glass and painted wood. This hand pad from 3M and Grainger provides an effective cleaning solution.