Learn More
Grainger Workbench Laminate 96 W 36 D
Catalog No. 502767838
Encompass_Preferred
The Workbench Top Laminate from Grainger is a durable and attractive work surface for garage, shop, or industrial use. It is constructed with a 1-1/4 inch thick particleboard core and covered in a chemical-resistant melamine laminate finish available in gray or brown. The workbench top measures 96 inches long by 24 inches wide, providing a spacious work area. It can support up to 2500 lbs when properly installed. The laminate surface resists stains, abrasions, and impacts. This workbench top requires assembly and mounting hardware for installation. It's an ideal durable work surface for a home garage, workshop, or industrial facility.
Catalog No. 50-276-7838 Supplier Grainger Supplier No. 16Y079
The Workbench Top Laminate from Grainger is a durable and attractive work surface for garage, shop, or industrial use. It is constructed with a 1-1/4 inch thick particleboard core and covered in a chemical-resistant melamine laminate finish available in gray or brown. The workbench top measures 96 inches long by 24 inches wide, providing a spacious work area. It can support up to 2500 lbs when properly installed. The laminate surface resists stains, abrasions, and impacts. This workbench top requires assembly and mounting hardware for installation. It's an ideal durable work surface for a home garage, workshop, or industrial facility.